Skip to Content
Merck
All Photos(3)

Key Documents

381985

Sigma-Aldrich

Nysted Reagent

20 wt. % suspension in THF

Synonym(s):

cyclo-Dibromodi-μ-methylene[μ-(tetrahydrofuran)]trizinc

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€147.00
1 KG
€962.00

€147.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€147.00
1 KG
€962.00

About This Item

Empirical Formula (Hill Notation):
C6H12Br2OZn3
CAS Number:
Molecular Weight:
456.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€147.00


Available to ship on17 April 2025Details


Request a Bulk Order

form

slurry

reaction suitability

reaction type: C-C Bond Formation

concentration

20 wt. % suspension in THF

density

1.186 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

C1CCOC1.Br[Zn]C[Zn]C[Zn]Br

InChI

1S/C4H8O.2CH2.2BrH.3Zn/c1-2-4-5-3-1;;;;;;;/h1-4H2;2*1H2;2*1H;;;/q;;;;;;2*+1/p-2

InChI key

CCTHTLJWXPUNGT-UHFFFAOYSA-L

Application

Reactant for:
  • Synthesis of amphidinolide T3 using a ring-closing metathesis and asymmetric dihydroxylation strategy
  • Methylenation of carbonyl compounds
  • Olefination reactions
Useful reagent for the methylenation of ketones and α-ketols.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F

Flash Point(C)

-26 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service