Skip to Content
Merck
All Photos(1)

Key Documents

378089

Sigma-Aldrich

Poly(tetrafluoroethylene oxide-co-difluoromethylene oxide) α,ω-diisocyanate

average Mn ~3,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NCO)NHCO2(CF2CF2O)x(CF2O)yCONHC6H3(NCO)CH3
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~3,000

refractive index

n20/D 1.352 (lit.)

viscosity

7000 cSt(20 °C)(lit.)

density

1.788 g/mL at 25 °C (lit.)

SMILES string

OC(O)(F)F.FC1OC1F.Cc2ccc(NC(O)=O)cc2N=C=O

General description

Ethylene to methylene ratio 1:1

Application

Reacts with alcohols and acids to form urethanes and amides, with lower coefficient of friction, wear and Tg.

Physical form

Aromatic diisocyanate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service