Skip to Content
Merck
All Photos(1)

Key Documents

376388

Sigma-Aldrich

(R)-(+)-Pulegone

≥90%

Synonym(s):

(+)-Pulegone, (R)-2-Isopropylidene-5-methylcyclohexanone, (R)-p-Menth-4(8)-en-3-one, p-Menth-4(8)-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2040703
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

138 mmHg ( 25 °C)

Assay

≥90%

form

liquid

optical activity

[α]20/D +23.5°, neat

refractive index

n20/D 1.486 (lit.)

bp

224 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

C[C@@H]1CC\C(=C(\C)C)C(=O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-Pulegone, a monoterpene ketone found in the essential oil of pennyroyal,[1] can induce abortion.[2]

Application

(R)-(+)-Pulegone may be used as a starting material to synthesize:
  • (2S,4R,6R,8S)-2,4,8-trimethyl-1,7-dioxaspiro[5.5]undecane, a spiroketal[3]
  • (R)-(+)-citronellic acid, an intermediate to prepare leucine-d3[4]
  • (+)-fawcettidine, a lycopodium alkaloid[5]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of (R)-(+)-pulegone in F344 rats.
Chen LJ, et al.
Drug Metabolism and Disposition, 29(12), 1567-1577 (2001)
From (R)-(+)-pulegone to (2S,4R,6R,8S)-2,4,8-trimethyl-1,7-dioxaspiro[5.5]undecane-A unique spiroacetal from the insect Kingdom.
Tu YQ, et al.
Tetrahedron Asymmetry, 6(2), 397-400 (1995)
Total Synthesis of (+)-Fawcettidine.
Kozak JA and Dake GR.
Angewandte Chemie (International Edition in English), 47(22), 4221-4223 (2008)
Synthesis of (2S, 4S)-and (2S, 4R)-[5,5,5-2H3] leucine from (R)-pulegone.
Hill RK, et al.
Canadian Journal of Chemistry, 72(1), 110-113 (1994)
Charalabos D Kanakis et al.
Phytochemical analysis : PCA, 23(1), 34-43 (2011-05-28)
Mentha pulegium L. (pennyroyal) is one of the four most commercially important Mentha species, even it is not a cultivated plant. It can be abundantly located in the Iberian Peninsula and North African countries. In Greece it grows in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service