Skip to Content
Merck
All Photos(1)

Key Documents

374016

Sigma-Aldrich

N-Allylcyclopentylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9NHCH2CH=CH2
CAS Number:
Molecular Weight:
125.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

155-160 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

SMILES string

C=CCNC1CCCC1

InChI

1S/C8H15N/c1-2-7-9-8-5-3-4-6-8/h2,8-9H,1,3-7H2

InChI key

CTSIKBGUCQWRIM-UHFFFAOYSA-N

General description

N-Allylcyclopentylamine is an organic building block. Boiling point of N-allylcyclopentylamine has been reported.[1]

Application

N-Allylcyclopentylamine may be used in the preparation of N,N-cyclopentylallylurea.[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodocyclisation of N-allyl ureas; a route to imidazolin-2-ones.
Moody CJ, et al.
ARKIVOC (Gainesville, FL, United States), 698, 706--706- (2000)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 201-201 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service