Skip to Content
Merck

365165

Sigma-Aldrich

Sodium L-lactate

99%

Synonym(s):

(S)-2-Hydroxypropionic acid sodium salt, L-Lactic acid sodium salt, Sarcolactic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06
Beilstein:
4567087
EC Number:
MDL number:
UNSPSC Code:
12352103

Pricing and availability is not currently available.

Assay

99%

optical activity

[α]20/D −12°, c = 1 in H2O

mp

163-165 °C (lit.)

SMILES string

[Na+].C[C@H](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1

InChI key

NGSFWBMYFKHRBD-DKWTVANSSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Alessio Innocenti et al.
    Bioorganic & medicinal chemistry, 17(7), 2654-2657 (2009-03-20)
    The inhibition of the beta-carbonic anhydrases (CAs, EC 4.2.1.1) from the pathogenic fungi Cryptococcus neoformans (Can2) and Candida albicans (Nce103) with carboxylates such as the C1-C5 aliphatic carboxylates, oxalate, malonate, maleate, malate, pyruvate, lactate, citrate and some benzoates has been

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service