Skip to Content
Merck
All Photos(1)

Key Documents

36037

Sigma-Aldrich

2′-(Dimethylamino)-2-biphenylyl-palladium(II) chloride Dinorbornylphosphine complex

≥99.0% (C)

Synonym(s):

Chloro-[2′-(dimethylamino)-2-biphenylyl]-(dinorbornylphosphine)-palladium, SK-CC01-A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37ClNPPd
CAS Number:
Molecular Weight:
560.45
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (C)

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

160-170 °C (dec.) (lit.)

SMILES string

C1C[C@@H]2C[C@H]1CC2PC3C[C@@H]4CC[C@H]3C4.CN(C)c5ccccc5-c6ccccc6[Pd]Cl

InChI

1S/C14H14N.C14H23P.ClH.Pd/c1-15(2)14-11-7-6-10-13(14)12-8-4-3-5-9-12;1-3-11-5-9(1)7-13(11)15-14-8-10-2-4-12(14)6-10;;/h3-8,10-11H,1-2H3;9-15H,1-8H2;1H;/q;;;+1/p-1/t;9-,10+,11+,12-,13?,14?;;

InChI key

YMZCYRNVJBOOCT-SBADXZPPSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Highly reactive, air and moisture stable palladium catalyst for C-C and C-N coupling reactions, e.g. Suzuki coupling, Heck reaction etc

Legal Information

sold in collaboration with Solvias AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Indolese, A. Schnyder
Eur. Pat. Appl. (2001)
S.C. Stinson
Chemical and Engineering News, october 29, 23-23 (2001)
Jean-Philippe Ebran et al.
Journal of the American Chemical Society, 129(21), 6931-6942 (2007-05-04)
A catalyst system was identified which promotes the Heck coupling of nonactivated vinyl phosphates with electron deficient alkenes providing a new entry to diene products from simple and readily accessible starting materials. In contrast to our earlier work exploiting P(t-Bu)3
A new generation of air stable, highly active Pd complexes for C--C and C--N coupling reactions with aryl chlorides.
Anita Schnyder et al.
Angewandte Chemie (International ed. in English), 41(19), 3668-3671 (2002-10-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service