Skip to Content
Merck
All Photos(1)

Key Documents

34070

Sigma-Aldrich

tert-Butyl 2,4-dibromobutyrate

technical, ≥85% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14Br2O2
CAS Number:
Molecular Weight:
302.00
Beilstein:
4660530
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical

Assay

≥85% (GC)

form

liquid

density

1.562 g/mL at 20 °C (lit.)

functional group

bromo
ester

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)C(Br)CCBr

InChI

1S/C8H14Br2O2/c1-8(2,3)12-7(11)6(10)4-5-9/h6H,4-5H2,1-3H3

InChI key

IELBUWARMJVXDU-UHFFFAOYSA-N

Application

tert-Butyl 2,4-dibromobutyrate was employed as building block in reaction with primary amines to yield azetidine-2-carboxylates.[1][2][3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 46, 2997-2997 (1981)
H.H. Wassermann et al.
Tetrahedron Letters, 1089-1089 (1979)
The Journal of Organic Chemistry, 46, 2991-2991 (1981)
Tingting Shi et al.
Oncology reports, 44(2), 457-468 (2020-07-07)
Aspirin, a nonsteroidal anti‑inflammatory drug (NSAID), is known to inhibit cell proliferation in a variety of cancers. However, the underlying mechanism of this inhibition remains unknown. We investigated the effects of aspirin on hepatocellular carcinoma (HCC) cells using in vitro and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service