Skip to Content
Merck
All Photos(1)

Documents

336629

Sigma-Aldrich

3-(Phenylthio)acrylic acid, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH=CHCO2H
CAS Number:
Molecular Weight:
180.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

102-106 °C (lit.)

SMILES string

OC(=O)\C=C/Sc1ccccc1

InChI

1S/C9H8O2S/c10-9(11)6-7-12-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6-

InChI key

QCLSYKCZWZYPIX-SREVYHEPSA-N

General description

3-(Phenylthio)acrylic acid is also referred as cis-β-phenyl thioacrylic acid. Ionization constants of cis-β-phenyl thioacrylic acid and their ring derivatives have been evaluated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transmission of Substituent Effects through Oxygen and Sulfur Atoms. III. The Ionization Equilibrium Constants of Ringsubstituted cis-jS-Phenoxyacrylic Acids.
Kajimoto O, et al.
Bulletin of the Chemical Society of Japan, 46(5), 1425-1428 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service