Skip to Content
Merck
All Photos(1)

Key Documents

332011

Sigma-Aldrich

4-Acetyl-4-phenylpiperidine hydrochloride

98%

Synonym(s):

1-(4-Phenyl-4-piperidinyl)-ethanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO · HCl
CAS Number:
Molecular Weight:
239.74
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

232-234 °C (lit.)

solubility

water: soluble(lit.)

SMILES string

Cl[H].CC(=O)C1(CCNCC1)c2ccccc2

InChI

1S/C13H17NO.ClH/c1-11(15)13(7-9-14-10-8-13)12-5-3-2-4-6-12;/h2-6,14H,7-10H2,1H3;1H

InChI key

JYDHZOIDIWUHDB-UHFFFAOYSA-N

General description

4-Acetyl-4-phenylpiperidine hydrochloride generates acetylperoxyl radical via laser flash photolysis in the presence of oxygen.

Application

Reactant for synthesis of neurotransmitter transport inhibitors with activity at dopamine receptor sites

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laser flash photolysis of new water-soluble peroxyl radical precursor.
El-Agamey A.
Journal of Photochemistry and Photobiology A: Chemistry, 203(1), 13-17 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service