Skip to Content
Merck
All Photos(3)

Documents

318108

Sigma-Aldrich

Chromium(III) acetate hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)7Cr3(OH)2
CAS Number:
Molecular Weight:
603.31
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Cr, 23.0-25.0% titration (by sodium thiosulfate)

reaction suitability

core: chromium

SMILES string

CC(=O)O[Cr](O)O.CC(=O)O[Cr](OC(C)=O)OC(C)=O.CC(=O)O[Cr](OC(C)=O)OC(C)=O

InChI

1S/7C2H4O2.3Cr.2H2O/c7*1-2(3)4;;;;;/h7*1H3,(H,3,4);;;;2*1H2/q;;;;;;;3*+3;;/p-9

InChI key

KDYFYAUIXHZPBG-UHFFFAOYSA-E

Application

Chromium(III) acetate hydroxide can be used for a variety of industrial applications which include dyeing, photographic emulsion, and tanning. It can also be used as a catalyst in the synthesis of monoacylglycerols by the glycidol-fatty acid reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CHROMIUM (III) ACETATE HYDROXIDE-AN EFFICIENT CATALYST FOR PREPARATION OF 1-MONOACYLGLYCEROLS BY THE GLYCIDOL-FATTY ACID REACTION
Janivs R, et al.
Journal of Food Lipids, 13(2), 199-209 (2006)
Polynuclear chromium (III) carboxlylates: Part 2. Chromium (III) acetate?what's in it?
Eshel M and Bino A
Inorgorganica Chimica Acta, 320(1-2), 127-132 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service