308242
Pyridoxine
98%
Synonym(s):
Pyridoxol, Vitamin B6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H11NO3
CAS Number:
Molecular Weight:
169.18
EC Number:
MDL number:
UNSPSC Code:
12162002
Recommended Products
Assay
98%
mp
159-162 °C (lit.)
SMILES string
Cc1ncc(CO)c(CO)c1O
InChI
1S/C8H11NO3/c1-5-8(12)7(4-11)6(3-10)2-9-5/h2,10-12H,3-4H2,1H3
InChI key
LXNHXLLTXMVWPM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
G Tolis et al.
The Journal of clinical endocrinology and metabolism, 44(6), 1197-1199 (1977-06-01)
The acute effect of pyridoxine (B6) on serum GH and PRL levels and its chronic effects on galactorrhea in nine subjects (group I, n=4, idiopathic galactorrhea with normal PRL levels and normal menses; Group II, n=5, galactorrhea-amenorrhea with increased PRL
Pyridoxine-dependent and pyridoxine-responsive seizures.
P Baxter
Developmental medicine and child neurology, 43(6), 416-420 (2001-06-21)
B Plecko et al.
Annals of neurology, 48(1), 121-125 (2000-07-14)
Diagnosis of pyridoxine-dependent epilepsy is based on the clinical response to high-dosage application of pyridoxine. Here, we report on 2 patients with pyridoxine-dependent epilepsy with significant elevation of pipecolic acid concentrations in plasma and cerebrospinal fluid (CSF) and further increase
J J van der Watt et al.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease, 15(6), 722-728 (2011-04-12)
Tuberculosis (TB) is increasing in incidence in certain parts of the world, particularly where there is a co-epidemic of human immunodeficiency virus/acquired immune-deficiency syndrome (HIV/AIDS), and it is associated with a significant degree of morbidity and mortality. One of the
Alex Soriano et al.
Antimicrobial agents and chemotherapy, 51(7), 2559-2563 (2007-05-02)
Hematological disturbances that develop during linezolid treatment are a major concern when linezolid is administered for prolonged periods of time. The aim of this study was to evaluate the influences of pyridoxine, rifampin, and renal function on hematological adverse events.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service