Skip to Content
Merck
All Photos(2)

Key Documents

297712

Sigma-Aldrich

4-Bromo-3-(trifluoromethyl)aniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CF3)NH2
CAS Number:
Molecular Weight:
240.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

81-84 °C/0.5 mmHg (lit.)

mp

47-49 °C (lit.)

functional group

bromo
fluoro

SMILES string

Nc1ccc(Br)c(c1)C(F)(F)F

InChI

1S/C7H5BrF3N/c8-6-2-1-4(12)3-5(6)7(9,10)11/h1-3H,12H2

InChI key

YGNISOAUPSJDJE-UHFFFAOYSA-N

Application

4-Bromo-3-(trifluoromethyl)aniline has been used in the preparation of:
  • 2,5-dibromo-(trifluoromethyl)benzene and 2-bromo-5-iodo-(trifluoromethyl)benzene[1]
  • AUY954, an aminocarboxylate analog of FTY720, a low nanomolar, monoselective agonist of the sphingosine-1-phosphate receptor[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 2 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shifeng Pan et al.
Chemistry & biology, 13(11), 1227-1234 (2006-11-23)
FTY720 is an immunomodulator with demonstrated efficacy in a phase II trial of relapsing multiple sclerosis. FTY720-phosphate, the active metabolite generated upon phosphorylation in vivo, acts as a potent agonist on four of the five known sphingosine-1-phosphate (S1P(1)) receptors. AUY954
Some Dihalo-(trifluoromethyl)-benzenes.
McBee ET, et al.
Journal of the American Chemical Society, 73(8), 3932-3934 (1951)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service