Skip to Content
Merck
All Photos(1)

Key Documents

295701

Sigma-Aldrich

Sulfur hexafluoride

≥99.75%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SF6
CAS Number:
Molecular Weight:
146.06
EC Number:
MDL number:
UNSPSC Code:
51442006
PubChem Substance ID:

vapor density

5.11 (vs air)

Quality Level

vapor pressure

22 mmHg ( 21.1 °C)

Assay

≥99.75%

bp

−64 °C/1 mmHg (lit.)

mp

−50 °C (lit.)

SMILES string

FS(F)(F)(F)(F)F

InChI

1S/F6S/c1-7(2,3,4,5)6

InChI key

SFZCNBIFKDRMGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfur hexafluoride is a stable gas with higher dielectric strength and density than air. It acts as an insulator in high-voltage circuit breakers as it shows high arc-quenching ability.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Tj Schütten et al.
JCI insight, 5(6) (2020-02-29)
BACKGROUNDSalt-sensitive hypertension is often accompanied by insulin resistance in obese individuals, but the underlying mechanisms are obscure. Microvascular function is known to affect both salt sensitivity of blood pressure and metabolic insulin sensitivity. We hypothesized that excessive salt intake increases
Sumit P Shah et al.
Retina (Philadelphia, Pa.), 33(2), 356-362 (2013-01-25)
To use spectral domain optical coherence tomography-guided duration of facedown positioning to study anatomical macular hole closure rates. Retrospective review of patients with macular holes undergoing 23-gauge pars plana vitrectomy and intraocular gas tamponade. Spectral domain optical coherence tomography imaging
Christoph Grouls et al.
Radiology, 267(2), 487-495 (2013-01-31)
To investigate the ability of vascular endothelial growth factor receptor type 2 (VEGFR2)-targeted ultrasonographic (US) microbubbles for the assessment of liver dysplasia in transgenic mice. Animal experiments were approved by the governmental review committee. Nuclear factor-κB essential modulator knock-out mice
Dana Gourevich et al.
Ultrasound in medicine & biology, 39(3), 507-514 (2013-01-22)
Ultrasound-mediated targeted drug delivery has been a subject for a dedicated research activity for several decades. Nevertheless, in vitro studies in this field of research are characterized by their inconsistencies. To improve the repeatability of such experiments, a novel approach of
Xinxiao Gao et al.
American journal of ophthalmology, 155(6), 1021-1027 (2013-03-26)
To identify risk factors for development of secondary full-thickness macular holes after pars plana vitrectomy with internal limiting membrane (ILM) peeling for myopic foveoschisis. Retrospective, interventional case series. We retrospectively reviewed the records of 42 eyes (42 patients) treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service