Skip to Content
Merck
All Photos(1)

Documents

291277

Sigma-Aldrich

Undecanoic δ-lactone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.459 (lit.)

bp

152-155 °C/10.5 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC1CCCC(=O)O1

InChI

1S/C11H20O2/c1-2-3-4-5-7-10-8-6-9-11(12)13-10/h10H,2-9H2,1H3

InChI key

YZRXRLLRSPQHDK-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Undecanoic δ-lactone has been found to be best substarte for good substrates for lactonase from Sulfolobus islandicus (SisLac).

Application

Undecanoic δ-lactone has been used as substrate to investigate the activities of stimulated and nonstimulated human serum paraoxonase (PON1) samples.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

312.8 °F

Flash Point(C)

156 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Hiblot et al.
PloS one, 7(10), e47028-e47028 (2012-10-17)
A new member of the Phosphotriesterase-Like Lactonases (PLL) family from the hyperthermophilic archeon Sulfolobus islandicus (SisLac) has been characterized. SisLac is a native lactonase that exhibits a high promiscuous phosphotriesterase activity. SisLac thus represents a promising target for engineering studies
S Billecke et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(11), 1335-1342 (2000-10-19)
It is well established that human serum paraoxonase (PON1) catalyzes the hydrolysis of organophosphate insecticides and nerve agents, as well as that of a number of aromatic carboxylic acid esters. Our laboratory has recently found a new class of PON1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service