Skip to Content
Merck
All Photos(1)

Key Documents

280666

Sigma-Aldrich

2-Ethoxyethyl methacrylate

contains 100 ppm hydroquinone monomethyl ether as inhibitor, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2OC2H5
CAS Number:
Molecular Weight:
158.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

contains

100 ppm hydroquinone monomethyl ether as inhibitor

refractive index

n20/D 1.429 (lit.)

bp

91-93 °C/35 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOCCOC(=O)C(C)=C

InChI

1S/C8H14O3/c1-4-10-5-6-11-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SFPNZPQIIAJXGL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vicente Compañ et al.
Biomaterials, 26(18), 3783-3791 (2005-01-01)
The oxygen permeability and diffusion coefficients of hydrogel membranes prepared with copolymers of 2-ethoxyethyl methacrylate (EEMA)/2,3-dihydroxypropylmethacrylate (MAG) with mole fraction of the second monomer in the range between 0 and 0.75 are described. Values of the permeability and diffusion coefficients
T V Chirila et al.
Journal of cataract and refractive surgery, 17(2), 154-162 (1991-03-01)
Development of improved hydrogels for soft intraocular lenses, based on 2-hydroxyethyl methacrylate monomer, requires the use of various other monomers and polymerization additives which have potential ocular toxicity. Three monomers, 2-hydroxyethyl methacrylate, methyl methacrylate, and 2-ethoxyethyl methacrylate, as well as
Dynamic simulation method to characterise oxygen transport in hydrogel membranes.
Zimei Rong et al.
Biomaterials, 27(23), 4266-4268 (2006-04-19)
B W Ziegelaar et al.
Journal of biomaterials science. Polymer edition, 9(8), 849-862 (1998-09-02)
We examined the regulation of collagenase production by the monocyte/macrophage THP-1 cell line when these cells were exposed to poly(2-hydroxyethyl methacrylate) (PHEMA) hydrogel surfaces with different chemistries and morphologies. Tissue culture modified polystyrene (TCP), used as a control surface, induced
K Tamareselvy et al.
Dental materials : official publication of the Academy of Dental Materials, 10(5), 290-297 (1994-09-01)
This research investigated the dynamic mechanical properties of two model dental restorative copolymers to elucidate the influence of pendent group length and addition of crosslinking agent. Two different monomethacrylate resins [(methyl methacrylate (MMA) or 2-ethoxyethylmethacrylate (EEM)] were added to various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service