Skip to Content
Merck
All Photos(2)

Key Documents

278076

Sigma-Aldrich

Tris(phenylthio)methane

98%

Synonym(s):

Triphenyl trithioorthoformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5S)3CH
CAS Number:
Molecular Weight:
340.53
Beilstein:
1914230
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

39-41 °C (lit.)

functional group

thioether

SMILES string

S(C(Sc1ccccc1)Sc2ccccc2)c3ccccc3

InChI

1S/C19H16S3/c1-4-10-16(11-5-1)20-19(21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15,19H

InChI key

YOQHDHPBEXTHCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris(phenylthio)methane has been used in the preparation of:
  • 1-phenylthio-tetrahydroisoquinolines via activated Pictet-Spengler cyclization[1]
  • homoallylchalcogenoacetals via reaction with allylsilanes in the presence of a Lewis acid[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thioorthoesters in the activated Pictet-Spengler cyclization. Synthesis of 1-thiosubstituted tetrahydroisoquinolines and carbon? carbon bond formation via sulfonyl iminium ions generated from N,S-sulfonyl acetals.
Silveira CC, et al.
Tetrahedron Letters, 44(32), 6137-6140 (2003)
Lewis acid-catalyzed coupling reactions of allylsilanes with tris (phenylchalcogeno) methane. Synthesis of homoallylchalcogenoacetals.
Silveira CC, et al.
Tetrahedron Letters, 37(34), 6085-6088 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service