Skip to Content
Merck
All Photos(2)

Documents

277029

Sigma-Aldrich

2-Chloro-2-methylbutane

98%

Synonym(s):

2-Methyl-2-chlorobutane, tert-Amyl chloride, tert-Pentyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2Cl
CAS Number:
Molecular Weight:
106.59
Beilstein:
1696930
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.405 (lit.)

bp

85-86 °C (lit.)

mp

−73 °C (lit.)

density

0.866 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)Cl

InChI

1S/C5H11Cl/c1-4-5(2,3)6/h4H2,1-3H3

InChI key

CRNIHJHMEQZAAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The influence of carbon-carbon multiple bonds on the solvolyses of 2-chloro-2-methylbutane has been critically evaluated through the extended Grunwald-Winstein equation. The rate constant for the SN 1 hydrolysis of 2-chloro-2-methylbutane has been measured near the consolute point of the liquid mixture of isobutyric acid and water.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina C Reis et al.
International journal of molecular sciences, 9(9), 1704-1716 (2009-03-28)
The influence of carbon-carbon multiple bonds on the solvolyses of 3-chloro-3-methylbutyne (1), 2-chloro-2-phenylpropane (2), 2-bromo-2-methyl-1-phenylpropane (3), 4-chloro-4-methyl-2-pentyne (4) and 2-chloro-2-methylbutane (5) is critically evaluated through the extended Grunwald-Winstein equation. Substrates 1, 3 and 5 lead to correlations with unexpected negative
Effect of the consolute point of isobutyric acid+ water on the rate of an SN1 hydrolysis reaction.
Kim YW and Baird JK.
International Journal of Thermophysics, 22(5), 1449-1461 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service