Skip to Content
Merck
All Photos(1)

Documents

263664

Sigma-Aldrich

3-(Chloromethyl)benzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C6H4COCl
CAS Number:
Molecular Weight:
189.04
Beilstein:
2436228
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.5748 (lit.)

bp

149-150 °C/20 mmHg (lit.)

density

1.33 g/mL at 25 °C (lit.)

SMILES string

ClCc1cccc(c1)C(Cl)=O

InChI

1S/C8H6Cl2O/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4H,5H2

InChI key

YCAIYRWHKSJKEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Chloromethyl)benzoyl chloride has been used in the preparation of:
  • new amphiphilic star polymers possessing a hyperbranched core and hydrophilic graft arms
  • water-soluble, solution-stable, bioactive prodrugs

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

International Journal of Pharmaceutics, 58, 143-143 (1990)
Synthesis of new amphiphilic star polymers derived from a hyperbranched macroinitiator by the cationic 'grafting from'method.
Weberskirch R, et al.
Macromolecular Chemistry and Physics, 200(4), 863-873 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service