Skip to Content
Merck
All Photos(1)

Documents

261637

Sigma-Aldrich

1-Naphthyl phosphate calcium salt trihydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C10H7OP(O)(OH)O]2Ca · 3H2O
CAS Number:
Molecular Weight:
540.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

192-193 °C (lit.)

storage temp.

−20°C

SMILES string

O.O.O.[Ca++].OP([O-])(=O)Oc1cccc2ccccc12.OP([O-])(=O)Oc3cccc4ccccc34

InChI

1S/2C10H9O4P.Ca.3H2O/c2*11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10;;;;/h2*1-7H,(H2,11,12,13);;3*1H2/q;;+2;;;/p-2

InChI key

ISLPOEWLFYKMBM-UHFFFAOYSA-L

Application

1-Naphthyl phosphate calcium salt trihydrate may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service