Skip to Content
Merck
All Photos(1)

Key Documents

259829

Sigma-Aldrich

cis-7-Dodecenyl acetate

Synonym(s):

Looplure, Z-7-DDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26O2
CAS Number:
Molecular Weight:
226.36
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

CCCC\C=C/CCCCCCOC(C)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter S Leal
Chemistry & biology, 11(8), 1029-1031 (2004-08-25)
The Asian elephant utilizes the same sex pheromone as a number of moth species, (Z)-7-dodecen-1-yl acetate encapsulated in a serum-derived albumin. The chemical signal is emitted in the urine and received in the mucus of the trunk. The unwrapping of
Insect pheromone in elephants.
L E Rasmussen et al.
Nature, 379(6567), 684-684 (1996-02-22)
L E Rasmussen et al.
Chemical senses, 22(4), 417-437 (1997-08-01)
In their natural ecosystems, adult male and female Asian elephants, Elephas maximus, live separately. For several weeks prior to ovulation, female elephants release a substance in their urine which elicits a high frequency of non-habituating chemosensory responses, especially flehmen responses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service