Skip to Content
Merck
All Photos(1)

Key Documents

254193

Sigma-Aldrich

Gallium(III) chloride

≥99.99%

Synonym(s):

Gallium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaCl3
CAS Number:
Molecular Weight:
176.08
EC Number:
MDL number:
UNSPSC Code:
12352302

vapor pressure

1 mmHg ( 48 °C)

Assay

≥99.99%

mp

78 °C (lit.)

density

2.47 g/mL at 25 °C (lit.)

SMILES string

Cl[Ga](Cl)Cl

InChI

1S/3ClH.Ga/h3*1H;/q;;;+3/p-3

InChI key

UPWPDUACHOATKO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Lower-temperature solvothermal techniques using GaCl3 and NaN3 or Li3N result in nanocrystalline GaN.
Radical mediator and catalyst in a number of organic bond formation reactions.

Packaging

product is packed in vials

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service