Skip to Content
Merck
All Photos(1)

Key Documents

252506

Sigma-Aldrich

9-Chloroanthracene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl
CAS Number:
Molecular Weight:
212.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

103-105 °C (lit.)

SMILES string

Clc1c2ccccc2cc3ccccc13

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Björklund et al.
Journal of environmental management, 159, 106-114 (2015-06-13)
Conventional stormwater treatment techniques such as sedimentation and filtration are inefficient for removing the dissolved and colloidal phases of hydrophobic organic compounds (HOCs) present in stormwater. Adsorption could be a promising technique for removing colloidal and dissolved pollutants. Five low-cost

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service