Skip to Content
Merck
All Photos(1)

Key Documents

249424

Sigma-Aldrich

2,3,4,5,6-Pentabromotoluene

99%

Synonym(s):

Pentabromotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Br5CH3
CAS Number:
Molecular Weight:
486.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

285-286 °C (lit.)

SMILES string

Cc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C7H3Br5/c1-2-3(8)5(10)7(12)6(11)4(2)9/h1H3

InChI key

OZHJEQVYCBTHJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delia Teresa Sponza et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 76(9-10), 2603-2622 (2017-11-24)
In this study, the degradations of 2,3,4,5,6-pentabromotoluene (PBT), 2,3,4,5,6-pentabromoethyl benzene (PBEB), triclosan (TCS) and gemfibrozil (GFZ) in raw hospital wastewater were investigated with cerium (IV) oxide and titanium (IV) oxide nanoparticles considering the mechanisms of adsorption, photolysis, and photocatalysis with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service