Skip to Content
Merck
All Photos(1)

Key Documents

248045

Sigma-Aldrich

3-Fluorophenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CO2H
CAS Number:
Molecular Weight:
154.14
Beilstein:
775898
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

42-44 °C (lit.)

SMILES string

OC(=O)Cc1cccc(F)c1

InChI

1S/C8H7FO2/c9-7-3-1-2-6(4-7)5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

YEAUYVGUXSZCFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adel Nefzi et al.
Bioorganic & medicinal chemistry letters, 19(17), 5169-5175 (2009-07-28)
Combinatorial chemistry offers a unique opportunity for the synthesis and screening of large numbers of compounds and significantly enhances the prospect of finding new drugs. Collaborative efforts with the Tuberculosis Antimicrobial Acquisition & Coordinating Facility (TAACF), have led to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service