Skip to Content
Merck
All Photos(1)

Key Documents

241075

Sigma-Aldrich

Allylamine

≥99%

Synonym(s):

2-Propen-1-ylamine, 3-Amino-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NH2
CAS Number:
Molecular Weight:
57.09
Beilstein:
635703
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2 (vs air)

Quality Level

vapor pressure

4.09 psi ( 20 °C)

Assay

≥99%

form

liquid

expl. lim.

22 %

refractive index

n20/D 1.420 (lit.)

bp

53 °C (lit.)

mp

−88 °C (lit.)

solubility

alcohol: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
water: miscible(lit.)

density

0.761 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCC=C

InChI

1S/C3H7N/c1-2-3-4/h2H,1,3-4H2

InChI key

VVJKKWFAADXIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allylamine was analyzed in the sevelamer hydrochloride (SVH) and sevelamer carbonate (SVC) drug substances by the capillary gas chromatography method, that uses a flame ionization detector[1].

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Raju V S N Kadiyala et al.
Scientia pharmaceutica, 82(1), 117-128 (2014-03-19)
A capillary gas chromatography method using a flame ionization detector has been developed for the trace analysis of allylamine (AA) in sevelamer hydrochloride (SVH) and sevelamer carbonate (SVC) drug substances. The method utilized a mega bore capillary column DB-CAM (30
Thomas Mayer-Gall et al.
Molecules (Basel, Switzerland), 24(17) (2019-08-29)
Poly- and cyclophosphazenes are excellent flame retardants but currently, are not used as textile finishing agents because water-soluble and permanent washing systems are missing. Here, we demonstrate for the first time, the successful usage of a water-soluble cyclotriphosphazene derivative for
Yonghang Xu et al.
Macromolecular rapid communications, 42(1), e2000446-e2000446 (2020-10-28)
Hydrogels and polydimethylsiloxane (PDMS) are complementary to each other, since the hydrophobic PDMS provides a more stable and rigid substrate, while the water-rich hydrogel possesses remarkable hydrophilicity, biocompatibility, and similarity to biological tissues. Herein a transparent and stretchable covalently bonded
Cheow Peng Ooi et al.
The Cochrane database of systematic reviews, 12, CD009361-CD009361 (2012-12-14)
Colesevelam is a second-generation bile acid sequestrant that has effects on both blood glucose and lipid levels. It provides a promising approach to glycaemic and lipid control simultaneously. To assess the effects of colesevelam for type 2 diabetes mellitus. Several
Galina Smushkin et al.
Diabetes, 62(4), 1094-1101 (2012-12-20)
We designed an experiment to examine the effect of bile acid sequestration with Colesevelam on fasting and postprandial glucose metabolism in type 2 diabetes. To do so, we tested the hypothesis that Colesevelam increases the disposition index (DI), and this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service