Skip to Content
Merck
All Photos(1)

Key Documents

235148

Sigma-Aldrich

Tris(trimethylsiloxy)ethylene

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€288.00

€288.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
10 G
€288.00

About This Item

Linear Formula:
[(CH3)3SiO]2C=CHOSi(CH3)3
CAS Number:
Molecular Weight:
292.59
Beilstein:
1962905
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€288.00


In StockDetails


Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.420 (lit.)

bp

54-56 °C/0.1 mmHg (lit.)

density

0.886 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)O\C=C(\O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C11H28O3Si3/c1-15(2,3)12-10-11(13-16(4,5)6)14-17(7,8)9/h10H,1-9H3

InChI key

FCZGHPGTZRTDNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris(trimethylsiloxy)ethylene has been used in:
  • stereospecific synthesis of insecticide ajuqarin-IV[1]
  • microwave-assisted synthesis of 2-hydroxy-1-phenylethanone[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multienzymatic preparation of 3-[(1R)-1-hydroxyethyl] benzoic acid and (2< i> S</i>)-hydroxy (phenyl) ethanoic acid.
Gennaro PD, et al.
Tetrahedron Asymmetry, 21(15), 1885-1889 (2010)
Stereospecific total synthesis of ajugarin-IV.
Kende AS, et al.
Tetrahedron Letters, 23(17), 1751-1754 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service