Skip to Content
Merck
All Photos(1)

Key Documents

234281

Sigma-Aldrich

Dimidium bromide

95%

Synonym(s):

3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide, Trypadine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18BrN3
CAS Number:
Molecular Weight:
380.28
Beilstein:
3809779
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

95%

mp

243-248 °C (lit.)

SMILES string

[Br-].C[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C20H17N3.BrH/c1-23-19-12-15(22)8-10-17(19)16-9-7-14(21)11-18(16)20(23)13-5-3-2-4-6-13;/h2-12,22H,21H2,1H3;1H

InChI key

MQOKYEROIFEEBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intercalating probe for nucleic acids.

replaced by

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Chemoprophylaxis of bovine trypanosomiasis T. congolense and T. vivax by phenathridinium S. 1553].
J DEOM
Annales de la Societe belge de medecine tropicale (1920), 30(1), 25-29 (1950-03-01)
T N Beliaeva et al.
Tsitologiia, 31(11), 1363-1368 (1989-11-01)
A natural DNA-intercalator plant benzo-c-phenanthridine alkaloid sanguinarine is more toxic for mouse transformed fibroblast L-cells in culture than synthetic DNA-intercalator ethidium bromide (EtB) and alkaloid berberine. Dimidium bromide is also an inhibitor of the L-cell growth. In assay conditions, growth
T P Mozhenok et al.
Tsitologiia, 40(6), 585-590 (1998-10-21)
Effect of DNA-intercalators ethidium bromide (EB, 0.005 and 0.015 mM) and dimidium bromide (DB, 0.005 and 0.010 mM) and antioxidative compounds acetylsalicylic acid (ASA, 0.05 and 0.50 mM) and beta-carotene (0.01, 0.02, 0.05 mM) on the phagosome-lysosome (P-L) fusion and
Dimidium bromide or phenanthridinium 1553; a note on the present position.
J CARMICHAEL
The Veterinary record, 62(17), 257-257 (1950-04-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service