Skip to Content
Merck
All Photos(2)

Key Documents

232858

Sigma-Aldrich

2-(2-Hydroxyethoxy)phenol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€112.00

€112.00

List price€160.00Save 30%

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€112.00

About This Item

Linear Formula:
HOCH2CH2OC6H4OH
CAS Number:
Molecular Weight:
154.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€112.00

List price€160.00Save 30%

Check Cart for Availability

Request a Bulk Order

Assay

98%

bp

128 °C/0.7 mmHg (lit.)

mp

99-100 °C (lit.)

functional group

hydroxyl

SMILES string

OCCOc1ccccc1O

InChI

1S/C8H10O3/c9-5-6-11-8-4-2-1-3-7(8)10/h1-4,9-10H,5-6H2

InChI key

AMCOCUDBDKVWRZ-UHFFFAOYSA-N

Application

2-(2-Hydroxyethoxy)phenol was used in the preparation of:
  • macrocyclic lipophilic benzocrown ethers[1]
  • coiled synthetic carboxylic ionophore with eight ether oxygens and five aromatic rings capable of encapsulating potassium ion[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, X-ray Crystal Studies and Metal Picrates Extraction Properties of Lipophilic Benzocrown Ethers.
Kowalska E, et al.
Australian Journal of Chemistry, 63(9), 1348-1357 (2010)
A coiled synthetic carboxylic ionophore capable of encapsulating a potassium ion.
Kuboniwa H, et al.
Journal of the Chemical Society. Chemical Communications, 21, 1468-1469 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service