Skip to Content
Merck
All Photos(1)

Key Documents

223042

Sigma-Aldrich

2,4,6-Tri-tert-butylnitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]3C6H2NO2
CAS Number:
Molecular Weight:
291.43
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

205-206 °C (lit.)

SMILES string

CC(C)(C)c1cc(c(c(c1)C(C)(C)C)[N+]([O-])=O)C(C)(C)C

InChI

1S/C18H29NO2/c1-16(2,3)12-10-13(17(4,5)6)15(19(20)21)14(11-12)18(7,8)9/h10-11H,1-9H3

InChI key

IMDZOFHRUMJNQR-UHFFFAOYSA-N

General description

The nitrogen-15 chemical shift (CS) tensor of 2,4,6-tri-tert-butylnitrobenzene was measured by solid-state NMR experiments[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn H Penner et al.
The Journal of organic chemistry, 68(11), 4258-4264 (2003-05-24)
Insight into the unexpectedly small range of isotropic nitrogen chemical shifts in nitrobenzene derivatives is gained through measurements of the chemical shift (CS) tensor by solid-state NMR experiments and ab initio molecular orbital (MO) and density functional theory (DFT) calculations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service