Skip to Content
Merck
All Photos(1)

Documents

219444

Sigma-Aldrich

o-Fluoro-DL-phenylalanine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
183.18
Beilstein:
3201186
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

99%

mp

243-246 °C (lit.)

SMILES string

NC(Cc1ccccc1F)C(O)=O

InChI

1S/C9H10FNO2/c10-7-4-2-1-3-6(7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)

InChI key

NYCRCTMDYITATC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Studying mechanism of P 450-mediated oxidation and rearrangement
  • Conversion of racemic α-arylalanines to (R)-β-arylalanines
  • Ribosomal translation of unnatural peptides
  • Synthesis of diisopropylpyridine acetamides for use as Kv1.5 potassium channel antagonists
  • Enantioselective hydrolysis of esters for resolution of nonprotein amino acids

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service