Skip to Content
Merck
All Photos(1)

Key Documents

207829

Sigma-Aldrich

Chromium(VI) oxide

≥99%

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

≥99%

mp

196 °C (dec.) (lit.)

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Vayssieres et al.
Journal of nanoscience and nanotechnology, 1(4), 385-388 (2003-08-14)
We are reporting here on the inexpensive fabrication and optical properties of an iron(III) oxide-chromium(III) oxide nanocomposite thin film of corundum crystal structure. Its novel and unique-designed architecture consists of uniformed, well-defined and oriented nanorods of Hematite (alpha-Fe2O3) of 50
M C García-Rodríguez et al.
Mutation research, 496(1-2), 145-151 (2001-09-12)
The effect of chlorophyllin on micronucleated polychromatic erythrocytes (MN-PCE) induction by chromium trioxide (CrO(3)) exposure in peripheral blood of mice was studied. Animals were treated with a single intraperitoneal dose of chlorophyllin (CHL) (20mg/kg), CrO(3) (20mg/kg), and CHL (20mg/kg) 4h
S Kondo et al.
European journal of biochemistry, 200(3), 689-698 (1991-09-15)
The chemical structure of the saccharide portion of Vibrio parahaemolyticus serotype 012 lipopolysaccharide was studied. Using chemical degradation and modification, as well as methylation analysis in combination with GLC-MS, laser-desorption mass spectrometry and 1H-NMR and 13C-NMR spectroscopy, the carbohydrate backbone
Adam Szelag et al.
Polish journal of pharmacology, 55(6), 1097-1103 (2004-01-20)
Hexavalent chromium compounds exhibit higher toxicity than its trivalent compounds since chromium ions in the +6 oxidation state easily cross biological membranes. It has recently been proposed that substances reducing chromium ions from the +6 to the less toxic +3
Seongmin Lee et al.
Organic letters, 11(1), 5-8 (2008-12-06)
The C14,15-dihydro-C22,25-epi north unit of cephalostatin 1 has been synthesized in 11 operations from commercially available hecogenin acetate via multiple reductions and oxidations. The key transformations include (i) Cr(VI)-catalyzed E-ring opening, (ii) C17 hydroxylation, and (iii) a base-triggered cyclization cascade.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service