Skip to Content
Merck
All Photos(1)

Documents

201898

Sigma-Aldrich

Direct yellow 50

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H24N6Na4O13S4
CAS Number:
Molecular Weight:
956.82
Colour Index Number:
29025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 40%

λmax

390 nm

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(NC(=O)Nc2ccc(N=Nc3cc(c4cccc(c4c3)S([O-])(=O)=O)S([O-])(=O)=O)c(C)c2)ccc1N=Nc5cc(c6cccc(c6c5)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C35H28N6O13S4.4Na/c1-19-13-21(9-11-29(19)40-38-23-15-27-25(33(17-23)57(49,50)51)5-3-7-31(27)55(43,44)45)36-35(42)37-22-10-12-30(20(2)14-22)41-39-24-16-28-26(34(18-24)58(52,53)54)6-4-8-32(28)56(46,47)48;;;;/h3-18H,1-2H3,(H2,36,37,42)(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4

InChI key

XRFRTDKENRGSSX-UHFFFAOYSA-J

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin-Lei Jia et al.
Plant reproduction, 34(2), 91-101 (2021-04-28)
The pollen coat, which forms on the pollen surface, consists of a lipid-protein matrix. It protects pollen from desiccation and is involved in adhesion, pollen-stigma recognition, and pollen hydration during interactions with the stigma. The classical methods used for pollen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service