Skip to Content
Merck
All Photos(2)

Key Documents

201464

Sigma-Aldrich

Variamine Blue RT Salt

Synonym(s):

4-Aminodiphenylamine diazonium sulfate, 4-Diazodiphenylamine sulfate, Azoic Diazo Component 22, Azoic Diazo No. 22

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€176.00

€176.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€176.00

About This Item

Linear Formula:
C6H5NHC6H4N2 · HSO4
CAS Number:
Molecular Weight:
293.30
Colour Index Number:
37240
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€176.00


Available to ship on14 April 2025Details


Request a Bulk Order

form

solid

solubility

1 M HCl: 10 mg/mL

λmax

377 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OS([O-])(=O)=O.N#[N+]c1ccc(Nc2ccccc2)cc1

InChI

1S/C12H10N3.H2O4S/c13-15-12-8-6-11(7-9-12)14-10-4-2-1-3-5-10;1-5(2,3)4/h1-9,14H;(H2,1,2,3,4)/q+1;/p-1

InChI key

FMRQRWPEQSPSDG-UHFFFAOYSA-M

Application

Variamine Blue RT Salt has been used as a dye in ultraviolet-visible spectrophotometric method based-uricase enzyme for the detection of uric acid in normal urine. It is also used in combination with naphthol AS-MX phosphate-variam to determine the cytochemical visualization of alkaline phosphatase activity.[1][2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Localization Mechanisms in Enzyme Cytochemistry
Studied with Alkaline Phosphatase-loaded
Erythrocyte Ghosts?
RAAP A K
The Journal of Histochemistry and Cytochemistry, 32, 815-820 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service