Skip to Content
Merck
All Photos(1)

Key Documents

198056

Sigma-Aldrich

3-Methylbenzophenone

99%

Synonym(s):

Phenyl m -tolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COC6H5
CAS Number:
Molecular Weight:
196.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.597 (lit.)

bp

183-185 °C/16 mmHg (lit.)

density

1.095 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)C(=O)c2ccccc2

InChI

1S/C14H12O/c1-11-6-5-9-13(10-11)14(15)12-7-3-2-4-8-12/h2-10H,1H3

InChI key

URBLVRAVOIVZFJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radka Pechancová et al.
Journal of separation science, 42(2), 556-565 (2018-11-10)
This work focused on the development and validation of the analytical procedure using gas chromatography equipped with vacuum-ultraviolet detection for the specific and sensitive determination of nine photoinitiators in food packages. Subsequently, a comparison of the combination of vacuum ultraviolet
Kanji Nagai et al.
Journal of chromatography. A, 1549, 85-92 (2018-03-28)
Poly(butylene terephthalate) based novel stationary phase (SP), composed of planar aromatic phenyl group together with ester group monomer units, was designed for supercritical fluid chromatography (SFC) use. As expected from its structure, this phase shows planarity recognition of isomeric aromatics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service