Skip to Content
Merck
All Photos(1)

Key Documents

191175

Sigma-Aldrich

DL-4-Chlorophenylalaninol

96%

Synonym(s):

(±)-2-(p-Chlorophenyl)-1-[hydroxymethyl)ethylamine, 2-Amino-3-(4-chlorophenyl)-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CH(NH2)CH2OH
CAS Number:
Molecular Weight:
185.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

solid

mp

80-82 °C (lit.)

SMILES string

NC(CO)Cc1ccc(Cl)cc1

InChI

1S/C9H12ClNO/c10-8-3-1-7(2-4-8)5-9(11)6-12/h1-4,9,12H,5-6,11H2

InChI key

RPHQMRWKOPBZQY-UHFFFAOYSA-N

Application

DL-4-Chlorophenylalaninol may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Long Chen et al.
Journal of the American Chemical Society, 130(51), 17222-17223 (2008-12-05)
The structures and dynamic solid-state behavior of four hydrogen-bonded layered metal-organic frameworks are discussed. Three of the complexes incorporate acetone guest molecules between layers; this is attributed to added flexibility in a framework ammonium component. One complex incorporates water guests

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service