Skip to Content
Merck
All Photos(1)

Key Documents

190497

Sigma-Aldrich

3′-Bromopropiophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COC2H5
CAS Number:
Molecular Weight:
213.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

39-41 °C (lit.)

SMILES string

CCC(=O)c1cccc(Br)c1

InChI

1S/C9H9BrO/c1-2-9(11)7-4-3-5-8(10)6-7/h3-6H,2H2,1H3

InChI key

QSHLXVTVXQTHBS-UHFFFAOYSA-N

Application

3′-Bromopropiophenone was used in the synthesis of o-, m- and p- isomers of (±)-2-[3-(hydroxybenzoyl)phenyl] propanoic acid. It was also used in the synthesis of 2-(methylamino)-1-(3-bromophenyl)propan-1-one.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Straightforward Syntheses of the Three Possible Isomers of (?)-2-[3-(Hydroxybenzoyl) phenyl] propanoic Acid, Metabolites of (?)-Ketoprofen.
Camps P and Farres X.
Synthetic Communications, 25(24), 3931-3938 (1995)
Novel aminopropiophenones as potential antidepressants.
Foley KF and Cozzi NV.
Drug Development Research, 60(4), 252-260 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service