Skip to Content
Merck
All Photos(1)

Key Documents

170852

Sigma-Aldrich

Hexyl nicotinate

97%

Synonym(s):

Nicotinic acid hexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
MDL number:

Assay

97%

SMILES string

CCCCCCOC(=O)c1cccnc1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Oestmann et al.
The British journal of dermatology, 128(2), 130-136 (1993-02-01)
This study assesses the variability of two non-invasive methods of measuring stratum corneum barrier function in vivo. Transepidermal water loss (TEWL), and the vascular response to hexyl nicotinate (HN) penetration as determined by laser-Doppler flowmetry, were measured in a group
K S Ryatt et al.
The British journal of dermatology, 119(3), 307-312 (1988-09-01)
The effect of short duration occlusion on skin penetration and stratum corneum water content was studied in vivo in eight human subjects. Percutaneous absorption of hexyl nicotinate was monitored non-invasively by laser Doppler velocimetry (LDV) following each of three randomly
Slaheddine Marrakchi et al.
Contact dermatitis, 55(1), 15-19 (2006-07-18)
Variation in human skin reactivity to various irritants in association with age and body region has been reported. Hexyl nicotinate (HN), a lipophilic nicotinate ester, was used to induce nonimmunologic contact urticaria in human volunteers of 2 age groups: 10
A P Lavrijsen et al.
The British journal of dermatology, 129(5), 547-553 (1993-11-01)
In this study, we characterized the stratum corneum barrier function in 39 patients with various keratinization disorders (autosomal dominant ichthyosis vulgaris [ADI] [n = 7], X-linked recessive ichthyosis [XRI] [n = 6], autosomal recessive congenital ichthyosis [CI] [n = 10]
L R Barclay et al.
Lipids, 32(3), 237-245 (1997-03-01)
The monoazaaromatics, pyridine (1), hexyl nicotinate (2), and quinoline (3) and diazaaromatics, pyrimidine (4) and purine (5), readily act as photo-initiators for the peroxidation of methyl linoleate in 0.50 M SDS at 37 degrees C giving free radical chain oxidations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service