Skip to Content
Merck
All Photos(1)

Key Documents

159565

Sigma-Aldrich

Diethyl 2-(2-cyanoethyl)malonate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CH2CH(CO2C2H5)2
CAS Number:
Molecular Weight:
213.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

115-118 °C/2 mmHg (lit.)

density

1.078 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(CCC#N)C(=O)OCC

InChI

1S/C10H15NO4/c1-3-14-9(12)8(6-5-7-11)10(13)15-4-2/h8H,3-6H2,1-2H3

InChI key

YJJLOESDBPRZIP-UHFFFAOYSA-N

General description

Diethyl 2-(2-cyanoethyl)malonate undergoes hydrothermal reaction with ZnCl2, NaN3 and NaOH to yield a new 1H-tetrazolate-5-butyrate coordination polymer.[1]

Application

Diethyl 2-(2-cyanoethyl)malonate was used in the synthesis of carbon-3-substituted 1,5,9-triazacyclododecanes, useful as RNA cleavage reagent for oligonucleotide tethering.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A diamond metal-organic framework with in situ generated 1H-tetrazolate-5-butyric acid ligand: Crystal structure, photoluminescence and high thermal stability.
Wu M-F, et al.
Inorganic Chemistry Communications, 14(1), 333-336 (2011)
J Hovinen
Bioconjugate chemistry, 9(1), 132-136 (1998-02-14)
Two derivatives of 1,5,9-triazacyclododecane bearing an alkylamino tether group at carbon-3 (7 and 13) were synthesized. The first of them, 7, was prepared starting from diethyl 2-(2-cyanoethyl)malonate and the second one, 13, from glycerol. In both cases, the cyclization reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service