Skip to Content
Merck
All Photos(1)

Key Documents

159360

Sigma-Aldrich

3-Fluorophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4NCO
CAS Number:
Molecular Weight:
137.11
Beilstein:
774879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.514 (lit.)

density

1.201 g/mL at 25 °C (lit.)

functional group

fluoro
isocyanate

storage temp.

2-8°C

SMILES string

Fc1cccc(c1)N=C=O

InChI

1S/C7H4FNO/c8-6-2-1-3-7(4-6)9-5-10/h1-4H

InChI key

RIKWVZGZRYDACA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Fluorophenyl isocyanate is an aryl fluorinated building block used in chemical synthesis.

Application

3-Fluorophenyl isocyanate was used in the synthesis of linezolid.[1] It was also used in the preparation of substituted 6-ureidopurines, 6(N2-3-fluorophenyl)ureidopurine.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of new substituted 6-ureidopurines and 6-ureido-9-(2, 3, 5-triacetylribofuranosyl) purines having cytokinin (plant growth promoting) activity.
Mhatre V and Joshi V.
Indian J. Chem. B, 41(12), 2667-2675 (2002)
New synthetic method of linezolid.
Yu D-S, et al.
Chinese Journal of Medicinal Chemistry / Zhong Guo Yao Wu Hua Xue Za Zhi, 2, 2-2 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service