Skip to Content
Merck
All Photos(1)

Key Documents

148326

Sigma-Aldrich

N,N′-Methylenebis(acrylamide)

electrophoresis grade, ≥99%

Synonym(s):

Bis-acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

electrophoresis grade

vapor density

5.31 (vs air)

Assay

≥99%

mp

>300 °C (lit.)

solubility

H2O: soluble 20 g/L at 20 °C

SMILES string

C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Methylenebis(acrylamide) is a cross linking agent that polymerizes with acrylamide and creates cross links within the polyacrylamide gel. It is capable of creating a network rather than linear chains which helps in maintaining the firmness of gel.

Application

N,N′-Methylenebis(acrylamide) has been used for synthesis of polymerized crystalline colloidal array (PCCAs). It has also been used for gel preparation.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy Olding et al.
Journal of medical physics, 36(1), 3-14 (2011-03-25)
This paper explores the combination of cone beam optical computed tomography with an N-isopropylacrylamide (NIPAM)-based polymer gel dosimeter for three-dimensional dose imaging of small field deliveries. Initial investigations indicate that cone beam optical imaging of polymer gels is complicated by
Roongnapa Suedee et al.
Journal of chromatography. A, 1114(2), 239-249 (2006-03-15)
A temperature sensitive dopamine-imprinted polymer was prepared in 80% aqueous methanol solution by free-radical cross-linking co-polymerisation of methacrylic acid and acrylamide at 60 degrees C in the presence of N,N-methylene-bis-acrylamide as the cross-linker and dopamine hydrochloride as template molecule. The
Jean Guglielminotti et al.
Anesthesiology, 122(5), 985-993 (2015-03-03)
Individual assessment of the amplitude of a physiologic reflex evoked by a standardized noxious test (SNT) before surgical stimulation has been suggested to predict movement upon the forthcoming surgical stimulation. This study aimed to compare the ability of pupillary dilatation
Yan Lu et al.
ACS nano, 4(12), 7078-7086 (2010-11-19)
Here, we demonstrate that bimetallic Au-Pt nanorods (NRs) can be grown in situ into thermosensitive core-shell microgel particles by a novel two-step approach. In the first step, Au NRs with an average width of 6.6 ± 0.3 nm and length
Bjørn Bjorvatn et al.
Sleep & breathing = Schlaf & Atmung, 19(4), 1387-1393 (2015-04-10)
The objective of the present study was to investigate the prevalence of insomnia and excessive sleepiness in relation to the presence and severity of obstructive sleep apnea (OSA). The sample consisted of patients referred to a university hospital on suspicion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service