Skip to Content
Merck
All Photos(2)

Documents

146234

Sigma-Aldrich

Barium 2-cyanoethylphosphate hydrate

Synonym(s):

2-Cyanoethyl phosphate barium salt hydrate, 3-(Phosphonooxy)propanenitrile barium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CH2OPO3Ba · xH2O
CAS Number:
Molecular Weight:
286.37 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: barium
reagent type: catalyst

SMILES string

[Ba++].[H]O[H].[O-]P([O-])(=O)OCCC#N

InChI

1S/C3H6NO4P.Ba.2H2O/c4-2-1-3-8-9(5,6)7;;;/h1,3H2,(H2,5,6,7);;2*1H2/q;+2;;/p-2

InChI key

SFFRVKYTJRRUNW-UHFFFAOYSA-L

Application

Reactant for NMR structural characterization of substrates bound to the active site of N-Acetylglucosaminyltransferase V

Other Notes

Contains barium acetate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service