Skip to Content
Merck
All Photos(1)

Key Documents

136921

Sigma-Aldrich

N,N-Dimethylbenzylamine

98%

Synonym(s):

N-Benzyldimethylamine, BDMA, DMBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)2
CAS Number:
Molecular Weight:
135.21
Beilstein:
1099620
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.501 (lit.)

bp

183-184 °C/765 mmHg (lit.)

mp

−75 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CN(C)Cc1ccccc1

InChI

1S/C9H13N/c1-10(2)8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3

InChI key

XXBDWLFCJWSEKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ballantyne et al.
Drug and chemical toxicology, 8(1-2), 43-56 (1985-01-01)
N-Benzyl-N,N-dimethylamine (BDMA), a polyester foam catalyst, was determined to have LD50 values of 0.65 (0.48-0.88) ml/kg perorally in the rat, and 1.66 (1.35-2.04) ml/kg by 24-hr occluded dermal contact in rabbits. The Lt50 for saturated vapor atmosphere exposure of rats
N Hayashi et al.
Organic letters, 3(14), 2249-2252 (2001-07-07)
[structure: see text] Three additional NMR databases, 1-3, in a chiral solvent are presented. The C.21-C.38 portion of oasomycin A is used to demonstrate the scope and limitation of the universal NMR database approach in a chiral solvent for assignment
B Ståhlbom et al.
International archives of occupational and environmental health, 70(6), 393-398 (1997-01-01)
The aim of the present study was to develop a method for generation of dimethylbenzylamine (DMBA) atmospheres in an exposure chamber and to investigate the possibility of using urinary DMBA metabolites for biological monitoring of exposure to DMBA. A DMBA
M A Patrauchan et al.
Journal of applied microbiology, 94(2), 266-272 (2003-01-22)
The aim of this work was to study the biodegradation of benzyldimethylalkylammonium chloride (BAC) by Aeromonas hydrophila sp. K, an organism isolated from polluted soil and capable of utilizing BAC as sole source of carbon and energy. High performance liquid
Y Kobayashi et al.
Organic letters, 3(14), 2253-2255 (2001-07-07)
[structure: see text] Examples of (13)C NMR desymmetrization of meso compounds are presented. On analysis of the NMR profiles of 1,3-diols, the additivity is recognized to predict the NMR profiles of 1,3,5-tirols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service