Skip to Content
Merck
All Photos(1)

Key Documents

135925

Sigma-Aldrich

2-Amino-3,5-dichloropyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4Cl2N2
CAS Number:
Molecular Weight:
163.00
Beilstein:
119376
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

81-83 °C (lit.)

SMILES string

Nc1ncc(Cl)cc1Cl

InChI

1S/C5H4Cl2N2/c6-3-1-4(7)5(8)9-2-3/h1-2H,(H2,8,9)

InChI key

OCWBGKZFOYMCCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-3,5-dichloropyridine was used in the synthesis of bis(2-amino-3,5-dihalopyridinium) tetrahalocuprate(II) compounds[1] and 2-fluoro-3,5-dichloropyridine[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New developments in the synthesis of lower fluorinated pyridines via diazotization-fluorination of aminopyridines in anhydrous hydrogen fluoride.
Boudakian, MM.
Journal of Fluorine Chemistry, 18(4), 497-506 (1981)
Grant W Tremelling et al.
Dalton transactions (Cambridge, England : 2003), (47)(47), 10518-10526 (2009-12-22)
A family of bis(2-amino-3,5-dihalopyridinium)tetrahalocuprate(II) compounds has been synthesized, including (3,5-diCAPH)2CuCl4 (1), (3,5-diCAPH)2CuBr4 (2), (3,5-diBAPH)2CuCl4 (3), and (3,5-diBAPH)2CuBr4 (4) [3,5-diCAPH = 2-amino-3,5-dichloropyridinium; 3,5-diBAPH = 2-amino-3,5-dibromopyridinium]. These complexes have been analyzed through single crystal X-ray diffraction and temperature dependent magnetic susceptibility. Compound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service