Skip to Content
Merck
All Photos(1)

Key Documents

124478

Sigma-Aldrich

N-Isopropylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH(CH3)2
CAS Number:
Molecular Weight:
135.21
Beilstein:
2205871
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.539 (lit.)

bp

55 °C/0.3 mmHg (lit.)

density

0.934 g/mL at 25 °C (lit.)

SMILES string

CC(C)Nc1ccccc1

InChI

1S/C9H13N/c1-8(2)10-9-6-4-3-5-7-9/h3-8,10H,1-2H3

InChI key

FRCFWPVMFJMNDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Isopropylaniline is an intermediate in propachlor degradation[1].

Application

N-Isopropylaniline has been used to study the mechanism of metabolism of propachlor by Pseudomonas sp.strain PEM1[1].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

189.9 °F - Cleveland open cup

Flash Point(C)

87.7 °C - Cleveland open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Propachlor and N-isopropylaniline residues in onions (Allium cepa) and muck soils.
B S Clegg et al.
Bulletin of environmental contamination and toxicology, 47(1), 104-111 (1991-07-01)
Sebastian R Sørensen et al.
Pest management science, 59(10), 1118-1124 (2003-10-17)
Mineralisation of the phenylurea herbicide isoproturon (3-(4-isopropylphenyl)-1,1-dimethylurea) and two of its known metabolites, 3-(4-isopropylphenyl)-1-methylurea (monodesmethyl-isoproturon) and 4-isopropylaniline, was studied in Danish agricultural soils with or without previous exposure to isoproturon. A potential for rapid mineralisation of isoproturon and the two
M Martin et al.
Applied and environmental microbiology, 65(2), 802-806 (1999-01-30)
Propachlor (2-chloro-N-isopropylacetanilide) is an acetamide herbicide used in preemergence. In this study, we isolated and characterized a soil bacterium, Acinetobacter strain BEM2, that was able to utilize this herbicide as the sole and limiting carbon source. Identification of the intermediates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service