Skip to Content
Merck
All Photos(1)

Documents

119180

Sigma-Aldrich

Methyl 2,5-dihydro-2,5-dimethoxy-2-furancarboxylate, mixture of cis and trans

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O5
CAS Number:
Molecular Weight:
188.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

refractive index

n20/D 1.449 (lit.)

bp

118-121 °C/12 mmHg (lit.)

density

1.175 g/mL at 25 °C (lit.)

SMILES string

COC1OC(OC)(C=C1)C(=O)OC

InChI

1S/C8H12O5/c1-10-6-4-5-8(12-3,13-6)7(9)11-2/h4-6H,1-3H3

InChI key

GHVXSSBZJZUDSO-UHFFFAOYSA-N

General description

Methyl 2,5-dihydro-2,5-dimethoxy-2-furancarboxylate is formed during electrochemical dimethoxylation of methyl-2-furoate in ceramic electrochemical microreactor.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Virginie Mengeaud et al.
Lab on a chip, 2(1), 39-44 (2004-04-22)
A ceramic electrochemical reactor (CEM) devoted to electrosyntheses was developed. The CEM was constituted by the assembly of five structured ceramic layers. On one layer, platinum interdigitated band electrodes with a submillimetre interelectrode gap were screen-printed. The microreactor chamber was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service