Skip to Content
Merck
All Photos(1)

Key Documents

107344

Carbon tetrachloride

99%

Synonym(s):

Tetrachloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl4
CAS Number:
Molecular Weight:
153.82
Beilstein:
1098295
EC Number:
MDL number:
PubChem Substance ID:

vapor density

5.32 (vs air)

vapor pressure

143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.460 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.594 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5

InChI key

VZGDMQKNWNREIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - Skin Sens. 1B - STOT RE 1 Inhalation

Target Organs

Liver,Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Søborg Pedersen et al.
Molecules (Basel, Switzerland), 25(5) (2020-03-07)
Despite promising anti-cancer properties in vitro, all titanium-based pharmaceuticals have failed in vivo. Likewise, no target-specific positron emission tomography (PET) tracer based on the radionuclide 45Ti has been developed, notwithstanding its excellent PET imaging properties. In this contribution, we present
J D Coombes et al.
Gut, 64(7), 1120-1131 (2014-06-07)
Chronic liver injury triggers a progenitor cell repair response, and liver fibrosis occurs when repair becomes deregulated. Previously, we reported that reactivation of the hedgehog pathway promotes fibrogenic liver repair. Osteopontin (OPN) is a hedgehog-target, and a cytokine that is
Samar Basu
Toxicology, 189(1-2), 113-127 (2003-06-25)
Hepatic injury through carbon tetrachloride (CCl(4)) induced lipid peroxidation is well known and has been extensively used in the experimental models to understand the cellular mechanisms behind oxidative damage and further to evaluate the therapeutic potential of drugs and dietary
Mark A Barnes et al.
Journal of leukocyte biology, 97(1), 161-169 (2014-11-16)
Recruitment of peripheral monocytes to the liver is a key contributor to the response to injury. MIF can act as a chemokine and cytokine, regulating innate immune responses in many tissues and cell types. We hypothesized that MIF contributes to
Xingbin Yang et al.
Food chemistry, 138(2-3), 1256-1264 (2013-02-16)
Water-soluble apple peel polysaccharides (APP) and apple flesh polysaccharides (AFP) were isolated from Pink Lady fruits, and their in vitro antioxidant capacities were characterised by DPPH(), HO(), and O(2)(-) systems, and ferric-reducing antioxidant power assay. Oral administration of APP at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service