Skip to Content
Merck
All Photos(1)

Key Documents

107328

Sigma-Aldrich

1-Benzoyl-4-piperidone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

bp

158-160 °C/0.2 mmHg (lit.)

mp

55-59 °C (lit.)

solubility

methylene chloride: soluble

SMILES string

O=C1CCN(CC1)C(=O)c2ccccc2

InChI

1S/C12H13NO2/c14-11-6-8-13(9-7-11)12(15)10-4-2-1-3-5-10/h1-5H,6-9H2

InChI key

NZAXGZYPZGEVBD-UHFFFAOYSA-N

Application

1-Benzoyl-4-piperidone can be used as starting reagent in the synthesis of fentanyl.

Biochem/physiol Actions

1-Benzoyl-4-piperidone and 1-methyl-4-piperidone reacts with triethyl phosphono-acetate in the presence of excess base and yields both the endocyclic and exocyclic olefins[1].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endocyclic vs exocyclic olefin formation from 4-piperidones via the wittig reaction.
Borne RF and Aboul-Enein HY.
Journal of Heterocyclic Chemistry, 9(4), 869-873 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service