Skip to Content
Merck
All Photos(3)

Key Documents

103268

Sigma-Aldrich

1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic anhydride

97%

Synonym(s):

Chlorendic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€67.50

€67.50


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€67.50

About This Item

Empirical Formula (Hill Notation):
C9H2Cl6O3
CAS Number:
Molecular Weight:
370.83
Beilstein:
92693
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€67.50


Available to ship on04 April 2025Details


Request a Bulk Order

vapor density

13 (vs air)

Assay

97%

form

solid

mp

235-239 °C (lit.)

SMILES string

ClC1=C(Cl)[C@@]2(Cl)C3C(C(=O)OC3=O)[C@]1(Cl)C2(Cl)Cl

InChI

1S/C9H2Cl6O3/c10-3-4(11)8(13)2-1(5(16)18-6(2)17)7(3,12)9(8,14)15/h1-2H/t1?,2?,7-,8+

InChI key

FLBJFXNAEMSXGL-SBUZQEQSSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service