Skip to Content
Merck

AABH99A49A11

AA BLOCKS, INC.

ethyl 2-(2-cyanoacetamido)-4,5-dimethylthiophene-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$108.00

$108.00


Estimated to ship onMay 10, 2025


To order products, please contact your local dealer.

Select a Size

Change View
1 G
$108.00

About This Item

CAS Number:
Molecular Weight:
266.32
MDL number:

$108.00


Estimated to ship onMay 10, 2025


To order products, please contact your local dealer.

Iupac Name

ethyl 2-[(cyanoacetyl)amino]-4,5-dimethyl-3-thiophenecarboxylate

Inchi Code

1S/C12H14N2O3S/c1-4-17-12(16)10-7(2)8(3)18-11(10)14-9(15)5-6-13/h4-5H2,1-3H3,(H,14,15)

InChI key

LYRZSPUDELNYMY-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

95%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/405924-25-6

Synonym

ethyl 2-(2-cyanoacetamido)-4,5-dimethylthiophene-3-carboxylate

MSDS

https://www.aablocks.com/downloadSds?cas=405924-25-6


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library