Skip to Content
Merck
All Photos(1)

Key Documents

AABH9616D2D2

AA BLOCKS, INC.

2-[(4-fluorophenyl)amino]nicotinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
213.21
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-(4-fluoroanilino)nicotinonitrile

Inchi Code

1S/C12H8FN3/c13-10-3-5-11(6-4-10)16-12-9(8-14)2-1-7-15-12/h1-7H,(H,15,16)

InChI key

CQPVCOCTBQRQIG-UHFFFAOYSA-N

storage temp.

2~8 C

Country of Origin

US

Product Link

https://www.aablocks.com/prod/874754-07-1

Synonym

2-(4-fluoroanilino)pyridine-3-carbonitrile

MSDS

https://www.aablocks.com/downloadSds?cas=874754-07-1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library